ENA388192255 Enamine

ethyl N-(2-amino-4-bromophenyl)carbamate

  • MDL number MFCD11198901
  • Cas Number : 1039915-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122577

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122577
Availability details for
Quantity
  • Availability
  • Add to Cart