ENA388192257 Enamine

(thiomorpholin-3-yl)methanol

  • MDL number MFCD02666403
  • Cas Number : 58729-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-91
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122579
Availability details for
Quantity
  • Availability
  • Add to Cart