ENA388192258 Enamine

1-methanesulfonylpropan-2-ol

  • MDL number MFCD01696728
  • Cas Number : 1977-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122580

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-122580
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart