ENA388192260 Enamine

4-[cyclopropyl(methyl)carbamoyl]benzoic acid

  • MDL number MFCD12173498
  • Cas Number : 1155169-16-6
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122582
Availability details for
Quantity
  • Availability
  • Add to Cart