ENA388192262 Enamine

2-(5-amino-1,3,4-thiadiazol-2-yl)ethan-1-ol

  • MDL number MFCD01203001
  • Cas Number : 904073-32-1
  • Molecular Weight : 145.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(5-amino-1,3,4-thiadiazol-2-yl)ethanol
Inchi Code 1S/C4H7N3OS/c5-4-7-6-3(9-4)1-2-8/h8H,1-2H2,(H2,5,7)
Inchi Key RBFBXGMHLCTYDB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-122590

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-122590
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart