ENA388192264 Enamine

(1-isocyanatoethyl)cyclopropane

  • MDL number MFCD18269677
  • Cas Number : 1339932-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122597
Availability details for
Quantity
  • Availability
  • Add to Cart