ENA388192277 Enamine

sodium 1-({[5-(4-nitrophenyl)furan-2-yl]methylidene}amino)-4-oxo-4,5-dihydro-1H-imidazol-2-olate pentahydrate

  • MDL number MFCD06198666

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122638

Safety Information

signalword Warning
safety_statement 22-24/25
msds https://www.enaminestore.com/catalog/EN300-122638
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart