ENA388192283 Enamine

methyl 4-bromo-5-cyclopropyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD16103387
  • Cas Number : 1291275-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122649
Availability details for
Quantity
  • Availability
  • Add to Cart