ENA388192287 Enamine

3-hydroxythiolane-3-carboxylic acid

  • MDL number MFCD12974321
  • Cas Number : 1172851-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122656

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122656
Availability details for
Quantity
  • Availability
  • Add to Cart