ENA388192288 Enamine

6-tert-butyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD11126888
  • Cas Number : 75413-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122657
Availability details for
Quantity
  • Availability
  • Add to Cart