ENA388192290 Enamine

4-methanesulfonyl-3-methylaniline

  • MDL number MFCD01463514
  • Cas Number : 1671-50-7
  • Molecular Weight : 185.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-156
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-122660

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-122660
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart