ENA388192296 Enamine

2-(tert-butoxy)-4-fluoroaniline

  • MDL number MFCD16744355
  • Cas Number : 1248597-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122667
Availability details for
Quantity
  • Availability
  • Add to Cart