ENA388192300 Enamine

(2,4-difluorophenyl)methanesulfonamide

  • MDL number MFCD09800677
  • Cas Number : 1001026-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122671

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122671
Availability details for
Quantity
  • Availability
  • Add to Cart