ENA388192302 Enamine

3,3'-dimethyl [1,1'-biphenyl]-3,3'-dicarboxylate

  • MDL number MFCD09032456
  • Cas Number : 1751-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122674
Availability details for
Quantity
  • Availability
  • Add to Cart