ENA388192304 Enamine

2-(5-chloro-1H-1,3-benzodiazol-2-yl)-6-methylphenol

  • MDL number MFCD11539903
  • Cas Number : 1094405-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122677

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122677
Availability details for
Quantity
  • Availability
  • Add to Cart