ENA388192305 Enamine

5-amino-1-(1,3-thiazol-2-yl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD20701752
  • Cas Number : 650638-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122682

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122682
Availability details for
Quantity
  • Availability
  • Add to Cart