ENA388192307 Enamine

ethyl 5-amino-1-(1,3-thiazol-2-yl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD22741308
  • Cas Number : 112267-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122684
Availability details for
Quantity
  • Availability
  • Add to Cart