ENA388192309 Enamine

1-[1-(2-chloro-4-fluorophenyl)cyclopropyl]methanamine

  • MDL number MFCD17282025
  • Cas Number : 1342004-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122686

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122686
Availability details for
Quantity
  • Availability
  • Add to Cart