ENA388192323 Enamine

3-(4-bromophenyl)-2-methylpropanoic acid

  • MDL number MFCD12152839
  • Cas Number : 66735-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122731

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122731
Availability details for
Quantity
  • Availability
  • Add to Cart