ENA388192327 Enamine

1-bromo-4-(2-isocyanatopropyl)benzene

  • MDL number MFCD11612998
  • Cas Number : 1082787-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122737

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122737
Availability details for
Quantity
  • Availability
  • Add to Cart