ENA388192328 Enamine

2-(6-methylpyridazin-3-yl)-1,2,3,4-tetrahydroisoquinolin-5-amine

  • MDL number MFCD13342686
  • Cas Number : 1307401-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122742

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122742
Availability details for
Quantity
  • Availability
  • Add to Cart