ENA388192330 Enamine

8-phenyl-8-azabicyclo[3.2.1]octan-3-one

  • MDL number MFCD01463740
  • Cas Number : 35193-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122744
Availability details for
Quantity
  • Availability
  • Add to Cart