ENA388192335 Enamine

ethyl 5-[hydroxy(phenyl)methyl]-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD19141573
  • Cas Number : 1275345-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120139
Availability details for
Quantity
  • Availability
  • Add to Cart