ENA388192336 Enamine

4-amino-3,5,6-trifluorobenzene-1,2-dicarbonitrile

  • MDL number MFCD00483216
  • Cas Number : 25693-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122751

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122751
Availability details for
Quantity
  • Availability
  • Add to Cart