ENA388192338 Enamine

2-chloro-6-phenoxybenzaldehyde

  • MDL number MFCD11156489
  • Cas Number : 181297-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122754
Availability details for
Quantity
  • Availability
  • Add to Cart