ENA388192344 Enamine

N-(2-aminoethyl)-4-methyl-2-oxo-2,3-dihydro-1,3-thiazole-5-sulfonamide hydrochloride

  • MDL number MFCD22741316
  • Cas Number : 1423034-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122834
Availability details for
Quantity
  • Availability
  • Add to Cart