ENA388192348 Enamine

4-methoxy-3-(propan-2-yl)benzoic acid

  • MDL number MFCD16303871
  • Cas Number : 33537-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122839
Availability details for
Quantity
  • Availability
  • Add to Cart