ENA388192352 Enamine

7-amino-1-methyl-1,2-dihydroquinolin-2-one

  • MDL number MFCD20697282
  • Cas Number : 58336-25-7
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-amino-1-methyl-2(1H)-quinolinone
Inchi Code 1S/C10H10N2O/c1-12-9-6-8(11)4-2-7(9)3-5-10(12)13/h2-6H,11H2,1H3
Inchi Key AVDMUZYDWIASIX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-122852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122852
Availability details for
Quantity
  • Availability
  • Add to Cart