ENA388192357 Enamine

thiomorpholine-3-carboxamide

  • MDL number MFCD02666349
  • Cas Number : 103742-31-0
  • Molecular Weight : 146.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-122866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122866
Availability details for
Quantity
  • Availability
  • Add to Cart