ENA388192360 Enamine

1-(2-fluorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD09043202
  • Cas Number : 926231-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122879

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122879
Availability details for
Quantity
  • Availability
  • Add to Cart