ENA388192362 Enamine

1-(3,5-dimethylbenzoyl)piperidine-2-carboxylic acid

  • MDL number MFCD09049183
  • Cas Number : 1101193-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122884

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122884
Availability details for
Quantity
  • Availability
  • Add to Cart