ENA388192363 Enamine

2-chloro-5-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD12922948
  • Cas Number : 99876-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-56
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122889
Availability details for
Quantity
  • Availability
  • Add to Cart