ENA388192370 Enamine

3-{[1-(2H-1,3-benzodioxol-5-yl)ethylidene]amino}-1-(oxolan-2-ylmethyl)thiourea

  • MDL number MFCD07270770
  • Cas Number : 571925-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122981

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122981
Availability details for
Quantity
  • Availability
  • Add to Cart