ENA388192382 Enamine

1-[(3-methyl-1,2-oxazol-5-yl)methyl]piperazine

  • MDL number MFCD08444202
  • Cas Number : 173850-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34769
Availability details for
Quantity
  • Availability
  • Add to Cart