ENA388192399 Enamine

7-chloro-2,3-dihydro-1H-indole

  • MDL number MFCD09040996
  • Cas Number : 114144-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-42
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-58142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58142
Availability details for
Quantity
  • Availability
  • Add to Cart