ENA388192413 Enamine

3-bromo-5-methoxyaniline

  • MDL number MFCD11878497
  • Cas Number : 16618-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39...41
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67112

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67112
Availability details for
Quantity
  • Availability
  • Add to Cart