ENA388192414 Enamine

5-bromo-2-chlorobenzene-1-sulfonamide

  • MDL number MFCD12026259
  • Cas Number : 1208077-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67485
Availability details for
Quantity
  • Availability
  • Add to Cart