ENA388192415 Enamine

2,3,4,5-tetrahydro-1H-2-benzazepine

  • MDL number MFCD09054850
  • Cas Number : 7216-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68684

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-68684
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart