ENA388192429 Enamine

methyl 2-bromo-3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoate

  • MDL number MFCD22741330
  • Cas Number : 95196-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83361
Availability details for
Quantity
  • Availability
  • Add to Cart