ENA388192436 Enamine

4-ethylcyclohexane-1-carbonitrile

  • MDL number MFCD11641480
  • Cas Number : 1096815-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90285
Availability details for
Quantity
  • Availability
  • Add to Cart