ENA388192437 Enamine

2-chloro-3-hydrazinyl-5,6-dimethylpyrazine

  • MDL number MFCD21262357
  • Cas Number : 1423034-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93148
Availability details for
Quantity
  • Availability
  • Add to Cart