ENA388192442 Enamine

2-amino-5-fluorobenzene-1-thiol

  • MDL number MFCD09909740
  • Cas Number : 33264-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97287

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97287
Availability details for
Quantity
  • Availability
  • Add to Cart