ENA388699576 Enamine

2-(2-chlorophenyl)cyclobutan-1-amine hydrochloride

  • MDL number MFCD23143994
  • Cas Number : 1427378-89-9
  • Molecular Weight : 218.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-chlorophenyl)cyclobutan-1-amine hydrochloride
Inchi Code 1S/C10H12ClN.ClH/c11-9-4-2-1-3-7(9)8-5-6-10(8)12;/h1-4,8,10H,5-6,12H2;1H
Inchi Key MMYQTGMGVYSVJD-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-118632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118632
Availability details for
Quantity
  • Availability
  • Add to Cart