ENA388699579 Enamine

2-methyl-4-(1-methyl-1H-pyrazol-4-yl)pyrimidine

  • MDL number MFCD23144001
  • Cas Number : 1427379-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119957
Availability details for
Quantity
  • Availability
  • Add to Cart