ENA388699584 Enamine

tert-butyl N-(2-aminoethyl)-N-(2-methoxyethyl)carbamate

  • MDL number MFCD23144009
  • Cas Number : 1427380-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120828

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-120828
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart