ENA388699586 Enamine

1-(1-methyl-1H-pyrazol-4-yl)-2-(pyrrolidin-2-yl)ethan-1-ol

  • MDL number MFCD23144014
  • Cas Number : 1427380-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121934
Availability details for
Quantity
  • Availability
  • Add to Cart