ENA388699593 Enamine

2-chloro-1-(3-cyclopropylmorpholin-4-yl)ethan-1-one

  • MDL number MFCD23144023
  • Cas Number : 1427379-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122533

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122533
Availability details for
Quantity
  • Availability
  • Add to Cart