ENA388699595 Enamine

3-cyclopropyl-1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

  • MDL number MFCD23144026
  • Cas Number : 1486485-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122714
Availability details for
Quantity
  • Availability
  • Add to Cart