ENA388699596 Enamine

2-(carbamoylmethoxy)-5-chlorobenzene-1-sulfonyl chloride

  • MDL number MFCD23144027
  • Cas Number : 1427379-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122716
Availability details for
Quantity
  • Availability
  • Add to Cart