ENA388699599 Enamine

3-bromo-5-methanesulfonyl-2-methylbenzoic acid

  • MDL number MFCD23144032
  • Cas Number : 1427379-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122892
Availability details for
Quantity
  • Availability
  • Add to Cart